missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Arabitol, 98%

Catalog No. AAA1310306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1310306 5 g
AAA1310314 25 g
AAA1310322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1310306 Supplier Thermo Scientific Chemicals Supplier No. A1310306

CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol

L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

Solubility
Soluble in water (50 mg/ml).

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7643-75-6
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064290
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
PubChem CID 439255
ChEBI CHEBI:18403
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 98°C to 103°C
Quantity 5 g
Beilstein 1720521
Sensitivity Hygroscopic
Merck Index 14,762
Solubility Information Soluble in water (50mg/ml).
Optical Rotation −11° (c=5 in 8% Borax Solution)
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material L-(-)-Arabitol
Hazard Category H500
EINECSNumber 231-582-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only