missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Arabitol, 98%

CAS: 7643-75-6 | C5H12O5 | 152.15 g/mol

Supplier:  Thermo Scientific Chemicals A1310314

Quantity 25 g

 View more versions of this product

Catalog No. AAA1310314


Only null left

Description

Description

L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

Solubility
Soluble in water (50 mg/ml).

Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

7643-75-6
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
439255
(2S,4S)-pentane-1,2,3,4,5-pentol
C5H12O5
MFCD00064290
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
CHEBI:18403
OCC(O)C(O)C(O)CO

Specifications

L-(-)-Arabitol
98°C to 103°C
25 g
1720521
14,762
Soluble in water (50mg/ml).
OCC(O)C(O)C(O)CO
(2S,4S)-pentane-1,2,3,4,5-pentol
439255
152.15
7643-75-6
C5H12O5
MFCD00064290
Hygroscopic
l-arabitol, l-arabinitol, l-arabinol, l-lyxitol, l---arabitol, arabinitol, l---arabinitol, arabinitol, l, 2s,4s-pentane-1,2,3,4,5-pentol, l--arabitol
HEBKCHPVOIAQTA-UHFFFAOYNA-N
−11° (c=5 in 8% Borax Solution)
152.15
CHEBI:18403
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-582-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions