missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine methyl ester hydrochloride, 99%

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol

$180.24 - $180.24

Identifiants chimiques

CAS 2491-20-5
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00063663
InChI Key IYUKFAFDFHZKPI-DOIAFFBONA-N
Synonym l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
PubChem CID 2733257
IUPAC Name methyl (2S)-2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](C)N
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC308360250
FDS Afficher les documents
Thermo Scientific Chemicals
308360250
25 g Glass bottle
chaque for $180.24
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

2491-20-5
139.58
IYUKFAFDFHZKPI-DOIAFFBONA-N
2733257
Cl.COC(=O)[C@H](C)N
C4H10ClNO2
MFCD00063663
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
methyl (2S)-2-aminopropanoate;hydrochloride

Spécifications

2491-20-5
White
98.5% min. (Argentometry)
C4H10ClNO2
MFCD00063663
+ 7.00
IYUKFAFDFHZKPI-DOIAFFBONA-N
methyl (2S)-2-aminopropanoate;hydrochloride
2733257
99%
L-Alanine methyl ester hydrochloride, 99%
109.0°C to 111.0°C
Authentic
Glass bottle
C3CH(NH2)CO2CH3·HCl
25 g
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
Cl.COC(=O)[C@H](C)N
139.58
139.58
Powder
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 219-652-4

SDS
Documents

Documents

RUO – Research Use Only