missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
Supplier: DWK Life Sciences 4225300250
Description
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
- Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
- Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
- Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
- Adjust spray pattern by varying the position of thumb on vent hole
- Autoclavable (after cap, loosening ring, and O-ring are removed)
Spécifications
| Autoclavable | |
| 250 mL | |
| Glass | |
| TLC Reagent Sprayer | |
| Standard TLC Reagents and Most Corrosive Liquids | |
| 24/40 | |
| 22.9 cm | |
| 8.5 cm |
| 8.45 oz. | |
| Screw thread ground joint, connecting cap, loosening ring | |
| 1 Case | |
| To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography | |
| 3 to 5 psi | |
| 9.01 in. | |
| 3.34 in. |