missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Keratinocyte SFM (1X), without calcium chloride
GREENER_CHOICE

Catalog No. 37010022
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37010022 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37010022 Supplier Gibco™ Supplier No. 37010022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keratinocyte-SFM (1X), without calcium chloride

Contains L-glutamine but no calcium chloride. Keratinocyte-SFM is supplied with prequalified human recombinant Epidermal Growth Factor 1-53 (EGF 1-53) and Bovine Pituitary Extract (BPE) in separate packaging. Components are not sold separately.

Specifications

Cell Type Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Calcium Chloride
Quantity 500 mL
Content And Storage • 500 mL Keratinocyte-SFM Basal Medium; store in refrigerator (2–8°C) and protect from light
• 25 mg Bovine Pituitary Extract (BPE); store in freezer (-5 to -20°C) and protect from light
• 2.5 μg EGF, Human Recombinant; store in freezer (-5 to -20°C) and protect from light

Complete Medium: store in refrigerator (2–8°C) and protect from light

Culture Type Mammalian Cell Culture
Show More Show Less
For how long can I store Keratinocyte-SFM at 4 degrees C?

The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C

For Research Use Only. Not for use in diagnostic procedures.