missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappler™ ProVent™ 7000 Isolation Gowns, Tie Closures Neck and Waist
Breathable, microporous fabric gowns offer general hazard protection for a variety of work environments from maintenance to general healthcare. Choose from elastic wrist or knit cuff options.
Supplier: Kappler™ PVS112WHXL2H
Description
Lightweight composite fabric utilizes microporous film technology to provides protection against hazardous particles and light liquid splash and maintain strength wet or dry
- Wraparound style opens to back
- Knit cuffs or elastic wrists
- Two neck ties, two waist ties
- Nonsterile
- White
Order Info
- Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front
Compliance
Meets ASTM™ F1670 and ASTM F1671 standards.
Spécifications
| General Purpose Gown | |
| None | |
| Wrap-around | |
| Microporous Coated Fabric | |
| Neck and Waist Ties | |
| Non-sterile | |
| PVS112WHXL2H | |
| Long Sleeve | |
| 17 lbs. |
| Maintenance, Spray Painting, Healthcare, Clinical | |
| Unisex | |
| X-Large | |
| White | |
| Single-use | |
| Knit | |
| Serged | |
| ASTM F1670, ASTM F1671 | |
| Not Autoclavable |