missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Ether (stabilized with BHT) 99.0+%, TCI America™

Supplier:  TCI America I0172500ML

Numéro de catalogue. I0172500ML


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Isopropyl Ether (stabilized with BHT)
-60°C
68°C
MFCD00008880
1159
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
2-propan-2-yloxypropane
7914
≥99.0% (GC)
108-20-3
Colorless
C6H14O
500 mL
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
CC(C)OC(C)C
102.177
102.18
Liquid

Identifiants chimiques

108-20-3
102.177
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
7914
CC(C)OC(C)C
C6H14O
MFCD00008880
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
2-propan-2-yloxypropane
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (2)-0362

missing translation for 'rtecsNumber' : TZ5425000

missing translation for 'tsca' : Yes

FDS
Documents

Documents

Promotions

Promotions