missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 97%

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

Supplier:  Thermo Scientific Chemicals A162310C

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA162310C


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

Solubility
Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, sodium and sodium oxides and potassium.
Spécifications

Spécifications

280°C to 285°C
37°C
1.8
Cl3FeH12O6
MFCD00149712
14,4019
Soluble in water,alcohol,glycerol,methanol,ether and acetone. Insoluble in ethyl acetate.
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
97%
Hygroscopic
Iron(III) chloride hexahydrate
10025-77-1
10,000 g
FeCl3·6H2O
UN3260
Ferric chloride hexahydrate
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
270.30 (162.21 Anhydrous)
Odorless
1.82

Identifiants chimiques

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
Cl3FeH12O6
MFCD00149712
Ferric chloride hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P333+P313-P362-P501c

H302-H315-H317-H318-H373

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-729-4

missing translation for 'rtecsNumber' : NO5425000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only