Learn More
CHEMetrics Iron Activator Solution
The Activator Solution is required for total iron analysis only.
Supplier:  CHEMetrics A6000
Numéro de catalogue. 13-299-217D
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Spécifications
Spécifications
Iron
Liquid
Iron Activator Solution
The Activator Solution is required for total iron analysis only.
6 x 10mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS