missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ PCR Clean-Up Kit

ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts.

$479.00 - $3,755.00

Spécifications

Content And Storage

The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
High-throughput Compatibility High-throughput Compatible
Product Line ChargeSwitch
Product Type PCR Clean-Up Kit
voir les spécifications

Comprend: ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Exige: Magnetic accessory


Produits 2
Numéro de catalogue Numéro du manufacturier. No. of Reactions Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. No. of Reactions Prix Quantity & Availability  
CS12000
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
CS12000
100 Preps
chaque for $479.00
Il en reste null
 
CS1200010
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
CS1200010
960 Preps
chaque for $3,755.00
Il en reste null
 
Description

Description

ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts (Figure 1). This kit is scalable for use on liquid-handling robots. This kit includes all the necessary reagents for the number of preps listed and requires the use of a magnetic accessory. Please see the Accessories Chapter (Chapter 6) to select the best magnetic accessory for your needs.

• Sample size: 25–50 μL
• High recovery of DNA fragments
• Rapid, one tube, 5 minute protocol

Order Info

Shipping Condition: Room Temperature

Spécifications

Spécifications

The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

High-throughput Compatible
PCR Clean-Up Kit
Room Temperature
Next-Generation Sequencing (NGS), Cloning, Standard PCR
PCR Products
Kit
ChargeSwitch
5 min
DNA from Enzymatic Reactions (e.g. PCR)
ChargeSwitch™ Chemistry
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.