Learn More
Intertek™ MIC Test Kit, Petroleum Microbiology Media: Nitrate Reducing Bacteria Media
Microbiology growth media products
Supplier: Intertek™ MK112A
Description
A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Nitrate Reducing Bacteria (NRB) can decrease corrosion by suppressing the growth of SRB, removing hydrogen sulfide and consuming hydrogen.
- NRB: for quantification of nitrate reducing bacteria
- NRB-Q: quick format single tube test for NRB
- The NRB-Q media provides a convenient way to estimate the concentration of NRB in water samples by changing the clarity of the media from clear to turbid/cloudy
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Spécifications
| NACE, TM0194-94 | |
| MIC Test Kit | |
| Tube |
| United States | |
| Cultivation of nitrate reducing bacteria, MIC testing | |
| 12 Tubes |