missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Insulin ELISA Kit

Catalog No. KAQ1251
Encompass
Change view
Click to view available options
Quantité:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
KAQ1251 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KAQ1251 Supplier Invitrogen™ Supplier No. KAQ1251
Saison des économies
View all promotions

missing translation for 'validMainframeMsg'

ELISA

The Human Insulin ELISA Kit quantifies human insulin in serum. The assay will recognize both the natural and recombinant insulin. Principle of the method The Human insulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.

Specifications

Numéro d'enregistrement P01308
Plage de l'essai 5.1-250 μlU/mL
Sensibilité d’essai 0.17 μIU/mL
Contenu et stockage Pre-coated plate(s), standard or calibrator, detector antibody(s), diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Conjugué HRP
Format de la trousse d’immunodosage Sandwich ELISA Kit
Type de produit ELISA
Famille de protéines Hormones & Receptors
Type d’échantillon Serum
Espèces cibles Human
À utiliser avec (application) Colorimetric Detection
À utiliser avec (équipement) Absorbance Microplate Reader
Alias du gène AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulinAA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin; IDDM; IDDM1; IDDM2; ILPR; IRDN; MODY10
ID du gène (Entrez) 3630
Symbole de gène INS
Temps d’incubation 0.75 hr.
CV intra-essai 8.5%, 5.4%
Contenus des kits Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Incubation Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Controls, Standard, Standard Dilution Buffer, Incubation Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Controls
Étiquette ou colorant Biotin
Marqueur génétique Insulin
Quantité 96 Tests
Statut réglementaire RUO
Volume d’échantillon Serum 50 μL
Shipping condition (Condition d'expédition) Wet Ice
Conditions de stockage 2-8°C
Cible Insulin
Durée du test 1 hr 20 min
Durée totale de l'essai 45 min
Show More Show Less