missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Solids pH Electrodes
A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).
$1093.85 - $1232.30
Spécifications
| Junction | Open |
|---|---|
| Material | Glass |
| pH Range | 1 to 11 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Internal Reference | ARGENTHAL |
| Numéro de catalogue | Numéro du manufacturier. | Connector Types | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Connector Types | Prix | Quantité | |||||
|
01-911-468
|
METTLER TOLEDO™
51343153 |
S7 |
chaque for $1,093.85
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
01-917-143
|
METTLER TOLEDO™
51344155 |
Multi-Pin |
chaque for $1,232.30
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
01-914-337
|
METTLER TOLEDO™
51343156 |
BNC IP67 |
chaque for $1,137.81
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Meats
- Cheese
- Seafood
- Dough
- Milk and cream
- Butter, yogurt, ice cream
- Fruits and vegetables
- Ready-made meals
- Solid and semi-solid food samples
Accessories & Care
- Open junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Spécifications
| Open | |
| 1 to 11 | |
| ARGENTHAL |
| Glass | |
| 0°C to 80°C | |
| LoT Glass |