missing translation for 'onlineSavingsMsg'
Learn More

Indium ICP Standard, 1000 ppm In in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PIN1KN500

 View more versions of this product

Catalog No. PIN1KN-500


Only null left

Description

Description

Specifications

Specifications

Indium ICP Standard
7440-74-6 , 7697-37-2 , 7732-18-5
90.0,8.1,0.09
1 mL = 1 mg In
In
Colorless
Approximately 0°C
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
500 mL
Natural Poly Bottle
1,000ppm In
93.0,8.43,0.09
Liquid
MFCD00134048
< 2
Standard
[In]
114.82

Chemical Identifiers

7440-74-6
114.82
APFVFJFRJDLVQX-UHFFFAOYSA-N
[In]
In
MFCD00134048
indium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions