missing translation for 'onlineSavingsMsg'
Learn More

Indium ICP Standard, 1000 ppm In in 3% HNO3, Ricca Chemical

$323.10 - $811.97

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
IUPAC Name indium
SMILES [In]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PIN1KN-100
View Documents
Ricca Chemical Company
PIN1KN100
100 mL Natural LDPE Bottle
Each for $323.10
Only null left
 
PIN1KN-500
View Documents
Ricca Chemical Company
PIN1KN500
500 mL Natural Poly Bottle
Each for $811.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7440-74-6
114.82
APFVFJFRJDLVQX-UHFFFAOYSA-N
[In]
In
MFCD00134048
indium

Specifications

Natural LDPE Bottle
1,000ppm In
93.0,8.43,0.09
Liquid
MFCD00134048
< 2
Standard
[In]
114.82
Indium ICP Standard
7440-74-6 , 7697-37-2 , 7732-18-5
90.0,8.1,0.09
1 mL = 1 mg In
In
Colorless
Approximately 0°C
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
100 mL
Videos
SDS
Documents

Documents