CAS RN 7440-74-6
Indium pieces, 99.99% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium rod, 2.38mm (0.094in) dia, 99.99% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium rod, 2.54mm (0.10 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.998% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium foil, 0.25mm (0.01in) thick, 99.99% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium ingot, 99.999% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium foil, 0.1mm (0.004in) thick, Puratronic™, 99.9975% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium wire, 2.0mm (0.08in) dia, Puratronic™, 99.998+% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium shot, 5mm (0.2in) & down, Puratronic™, 99.9999% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium shot, 4-8mm (0.2-0.3in), Puratronic™, 99.9999% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis)
CAS: 7440-74-6 Molecular Formula: In Molecular Weight (g/mol): 114.82 MDL Number: MFCD00134048 InChI Key: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC Name: indium SMILES: [In]