missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Imidazole, ACS, 99+%
Spécifications
Spécifications
| CAS | 288-32-4 |
| Melting Point | 88-92° |
| Density | 1.03 |
| Boiling Point | 256° |
| Flash Point | 145°(293°F) |
| Molecular Formula | C{3}H{4}N{2} |
| Quantity | 100 g |
| MDL Number | MFCD00005183 |
| UN Number | UN3263 |
| Merck Index | 14,4912 |
| Afficher plus de résultats |