missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 453360050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC453360050

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Spécifications

Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Authentic
C8H6N2O2·ClH
imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride, imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride 1:x
C1=CN2C=CN=C2C=C1C(=O)O.Cl
198.61
198.61
1423031-35-9
96.0 to max. 104% min. (Argentometry)
5g
OQGRMPZJJDBYBL-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-7-carboxylic acid;hydrochloride
71756722
97%

Identifiants chimiques

1423031-35-9
198.61
imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride, imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride 1:x
imidazo[1,2-a]pyridine-7-carboxylic acid;hydrochloride
C8H6N2O2·ClH
OQGRMPZJJDBYBL-UHFFFAOYSA-N
71756722
C1=CN2C=CN=C2C=C1C(=O)O.Cl
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only