missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-2 ProQuantum Immunoassay Kit
GREENER_CHOICE

Numéro de catalogue. A356035
Encompass
Modifier l'affichage
Click to view available options
Quantité:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A356035 5 x 96 Tests
A35603 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A356035 Fournisseur Invitrogen™ Code fournisseur A356035

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

ELISA

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.

Spécifications

Numéro d'enregistrement P60568
Plage de l'essai 0.0128-5000 pg/mL
Sensibilité d’essai <0.01 pg/mL
Contenu et stockage Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5 to -30°C.

Conjugué FAM
Méthode de détection Taqman™ Real-time PCR
Format de la trousse d’immunodosage Proximity-based Immunoassay
Type de produit ProQuantum
Type d’échantillon Plasma, Serum, Supernatant
Espèces cibles Human
À utiliser avec (application) Protein Quantitation
À utiliser avec (équipement) qPCR Instruments
Alias du gène aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGFaldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF; IL-2; TCGF; lymphokine
ID du gène (Entrez) 3558
Symbole de gène IL2
Temps d’incubation 1 hr.
Contenus des kits Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Étiquette ou colorant FAM™
Alias des protéines IL-2; T-cell growth factor; TCGF
Quantité 5 x 96 Tests
Statut réglementaire RUO
Volume d’échantillon Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Shipping condition (Condition d'expédition) Dry Ice
Conditions de stockage -20°C
Cible IL-2
Durée du test 30 min
Durée totale de l'essai 2 hr
Afficher plus de résultats Afficher moins de résultats