Learn More
Inorganic Ventures 1000ug mL OXALATE 125mL (Inorganic Ventures)
Supplier: Inorganic Ventures ICOXA1125ML
Matrix: Water (H2O), Starting Material: Sodium oxalate, Analyte: C2O4-2 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search