Learn More
Thermo Scientific™ HyperSep™ Javelin Direct-Connect Online SPE Columns
Clean-up and concentrate target analytes in minutes with these direct-connection SPE guard columns, featuring fingertight design for convenience and efficiency.
Supplier:  Thermo Scientific™ 60310302
 Afficher plus de versions de ce produit
Numéro de catalogue. 03-150-353
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep Javelin Direct-Connect Online SPE Columns feature a unique fingertight design for convenience and efficiency.
HyperSep Javelin Direct-Connect Online SPE Columns are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
Spécifications
Spécifications
HyperSep Retain-CX; 3.0 mm ID
Conventional HPLC System
HyperSep Retain CX
3.0 mm
Online Solid Phase Extraction
HyperSep
Online Solid Phase Extraction Column
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS