missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate, For 1L standard solution, 0.2 M HCl (0.2N), Honeywell Fluka™

for 1L standard solution, 0.2 M HCl (0.2N)

$191.00 - $996.43

Identifiants chimiques

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
6000769
Afficher les documents
Honeywell-Fluka
382871EA
1 Pc.
chaque for $191.00
Il en reste null
 
60046717
Afficher les documents
Honeywell-Fluka
382876X1EA
6 x 1 Ea.
paquet for $996.43
Il en reste null
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécifications

Identifiants chimiques

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Spécifications

7647-01-0
190 mmHg (25°C)
ClH
UN1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46g/mol
Ampoule
1 Pc.
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric acid concentrate
Videos
Sécurité et manipulation

Sécurité et manipulation

P305 + P351 + P338

missing translation for 'shelfLife' : 1260 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Hydrochloric acid

Recommended Storage : Room Temp

FDS
missing translation for 'documents'

missing translation for 'documents'