missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Certified, 20% (v/v) ±1%, LabChem™

Supplier:  LabChem LC150904

 Afficher plus de versions de ce produit

Numéro de catalogue. LC150904


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Hydrochloric Acid (1+4)
7647-01-0,7732-18-5
Passes Test
Traceable to NIST
4 L
Liquid
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
Certified
Colorless
91.5,8.5
Poly Bottle
Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
ClH
HCl
UN1789
Soluble in water
Cl
36.46
CHEBI:17883
20% ±1% (v/v)

Identifiants chimiques

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

Recommended Storage : Room Temperature

FDS
Documents

Documents

Promotions

Promotions