Learn More
Hydrochloric acid, 5% v/v aq. soln.
CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
$59.30 - $59.30
Identifiants chimiques
| CAS | 7647-10-1 |
|---|---|
| Molecular Formula | Cl2Pd |
| Molecular Weight (g/mol) | 177.32 |
| MDL Number | MFCD00003558 |
| InChI Key | PIBWKRNGBLPSSY-UHFFFAOYSA-L |
| Synonym | palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440 |
| PubChem CID | 24290 |
| ChEBI | CHEBI:53434 |
| SMILES | [Cl-].[Cl-].[Pd++] |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA43335K2
|
Thermo Scientific Chemicals
043335K2 |
1 L |
chaque for $59.30
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 7647-10-1 | |
| 177.32 | |
| PIBWKRNGBLPSSY-UHFFFAOYSA-L | |
| 24290 | |
| [Cl-].[Cl-].[Pd++] |
| Cl2Pd | |
| MFCD00003558 | |
| palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440 | |
| CHEBI:53434 |
Spécifications
| Colorless to Yellow | |
| 5% v/v aq. soln. | |
| Cl2Pd | |
| MFCD00003558 | |
| palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440 | |
| [Cl-].[Cl-].[Pd++] | |
| 177.32 | |
| CHEBI:53434 | |
| Hydrochloric acid |
| 7647-10-1 | |
| 1 L | |
| Liquid | |
| UN1789 | |
| PIBWKRNGBLPSSY-UHFFFAOYSA-L | |
| palladium(2+) dichloride | |
| 24290 | |
| 36.46 |
Sécurité et manipulation
P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
H290-H315-H318
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only