missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1 N, Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H12020LT

 Afficher plus de versions de ce produit

Numéro de catalogue. 18610861


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Hydrochloric Acid
0.100 N ± 0.001 N
Poly Bottle
Ungraded
7647-01-0,7732-18-5
99.64%,0.36%
20 L

Identifiants chimiques

7647-01-0
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.