missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4MAb Powdered Media

Catalog No. SH3053502
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3053502 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3053502 Supplier Cytiva Supplier No. SH30535.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increase process yields for manufacturing of human and humanized recombinant antibodies.

SFM4MAb is a low protein formulation and is optimized for downstream purification using protein A, protein G, and other matrices to facilitate product recovery. The medium is formulated using proprietary lipid and phospholipid complexing process for enhanced stability and growth promotion of various cell types.

SFM4MAb is available in liquid and powder formats in user-friendly packaging.

  • Designed for high cell yield and antibody production
  • Complexed lipids for enhanced stability
  • Allows for direct or sequential adaptation
  • Optimized for protein A, protein G, and other purification matrices
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

Specifications

Form Powder
Sterility Non-sterile
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture