missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4Insect Powdered Cell Culture Media

Catalog No. SH3091201
Change view
Click to view available options
Quantity:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3091201 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3091201 Supplier Cytiva Supplier No. SH30912.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ADCF (animal-derived component-free) medium supports recombinant protein production with multiple insect cell lines.

SFM4Insect medium provides excellent growth of many key insect cell lines while requiring minimal adaptation. Additionally, SFM4Insect medium has been successfully tested in a variety of culture systems up to and including high density bioreactors.

  • For use with the baculovirus expression vector system (BEVS)
  • Designed for high cell yield and recombinant protein production
  • Complexed lipids for enhanced stability
  • Supports multiple insect cell lines
  • Supports direct or sequential adaptation
  • Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP)

Specifications

Form Powder
Sterility Non-sterile
With Additives Glutamine
Without Additives No Sodium Bicarbonate
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture