missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Catalog No. SH3002802
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
1000 mL
6 x 500 mL
10 L
20 L
6 x 1000 mL
Concentration:
1 X
10 X
With Additives:
Calcium, Magnesium
None
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration With Additives Without Additives
SH3002802 500 mL 1 X None No Calcium, No Magnesium, No Phenol Red
SH3002801 100 mL 1 X None No Calcium, No Magnesium, No Phenol Red
SH30028LS 6 x 1000 mL 1 X None No Calcium, No Magnesium, No Phenol Red
SH3026402 1000 mL 1 X Calcium, Magnesium No Phenol Red
SH3002808 10 L 1 X None No Calcium, No Magnesium
SH30028FS 6 x 500 mL 1 X None No Calcium, No Magnesium, No Phenol Red
SH30264FS 6 x 500 mL 1 X Calcium, Magnesium No Phenol Red
SH3002803 1000 mL 1 X None No Calcium, No Magnesium, No Phenol Red
SH3026401 500 mL 1 X Calcium, Magnesium No Phenol Red
SH3037803 1 L 10 X None No Calcium, No Magnesium, No Phenol Red
SH3037802 500 mL 10 X None No Calcium, No Magnesium, No Phenol Red
SH3002804 20 L 1 X None No Calcium, No Magnesium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. SH3002802 Supplier Cytiva Supplier No. SH30028.02
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer cell culture with PBS in solution.

  • Format: Liquid

Specifications

Certifications/Compliance Manufactured under cGMP and ISO 9001 certified facilities
Content And Storage 15° to 30°C
Description HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red
Concentration 1 X
Form Liquid
Product Type Dulbecco's Phosphate Buffered Saline
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
With Additives None
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level ≤0.025EU/mL
For Use With (Application) Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
Osmolality 276±10mOsm/kg H2O
pH 7 to 7.6
Quantity 500 mL
Regulatory Status RUO
Show More Show Less