missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ AdvanceSTEM™ Human Somatic Stem Cell Media

Catalog No. SH3088902
Change view
Click to view available options
Product Type:
Adipogenic Differentiation Medium
Chondrogenic Differentiation Medium
Osteogenic Differentiation Medium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
SH3088902 Chondrogenic Differentiation Medium
SH3088102 Osteogenic Differentiation Medium
SH3088602 Adipogenic Differentiation Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3088902 Supplier Cytiva Supplier No. SH30889.02

Research mesenchymal stem cells with HyClone™ AdvanceSTEM™ Human Somatic Stem Cell Media.

Specifications

Cell Type Mesenchymal Stem Cells
Form Liquid
Product Type Chondrogenic Differentiation Medium
Sterility Sterile-filtered
Quantity 450 mL
Content And Storage Store at 2° to 8°C away from light