missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline, ES Cell Qualified

Catalog No. SH3085003
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3085003 1000 mL
SH3085002 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3085003 Supplier Cytiva Supplier No. SH30850.03
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Support growth of embryonic cell culture growth with HyClone™ AdvanceSTEM™ Dulbecco's Phosphate Buffered Saline.

Specifications

Certifications/Compliance cGMP and ISO 91:2 Guidelines
Content And Storage 15 to 30°C
Concentration 1 X
Product Type DPBS
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Calcium, No Magnesium
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.2
Product Line AdvanceSTEM ES Qualified
Quantity 1000 mL