missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HyClone™ ActiSM™ Cell Culture Media

Catalog No. SH3104002
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3104002 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3104002 Supplier Cytiva™ Supplier No. SH31040.02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A chemically defined ADCF cell culture medium optimized for high-yield protein production in CHO cell culture processes.

ActiSM™ cell culture media is a chemically defined animal-derived component-free (ADCF) cultured media optimized for high-yield protein production for bioprocessing. This HyClone™ medium is designed for use with ActiPro™ cell culture media.

  • Chemically defined ADCF formulation optimized for high-yield protein production
  • Does not contain hypoxanthine, thymidine, and glycine (HAT supplement)
  • Recommended for first step when adapting cells cultured in other lean cell culture media

ActiSM™ medium is intended for use in combination with ActiPro™ medium and Cell Boost™ 7a and Cell Boost™ 7b supplements. This HyClone™ medium is part of the ActiPro™ system of media and supplements designed with adaptation and production in mind. ActiSM™ and the ActiPro™ system are suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S. Once your process has been adapted for use with the ActiSM™ medium, we recommend utilizing ActiPro™ medium for production.

ActiSM™ cell culture medium is available in both powder and liquid formats. ActiSM™ medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring CHO batch-to-batch consistency. Cell culture media formulations and delivery systems can be customized to your specific process requirements or optimized to maximize process yields. Our cell culture media and development service is available to help you analyze, develop, and optimize your cell culture strategies.

Specifications

Cell Line CHO
Cell Type CHO
Classification Protein Production
Form Liquid
Product Type Cell Culture Media
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives Poloxamer 188
Without Additives No Glutamine, No Thymidine, No Hypoxanthine, No Glycine
Product Line HyClone ActiSM
Quantity 1000 mL
Culture Type Mammalian Cell Culture
Expression System Mammalian
Show More Show Less