missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Osteocrin ELISA Kit
ELISA
$890.00 - $4,420.00
Spécifications
| Accession Number | musclin; Ostc; Osteocrin; Ostn; Processed Osteocrin |
|---|---|
| Assay Range | 0.55 to 125 ng/mL |
| Assay Sensitivity | 0.55 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
EH355RBX5
|
Invitrogen™
EH355RBX5 |
5 x 96 Tests |
chaque for $4,420.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
EH355RB
|
Invitrogen™
EH355RB |
96 Tests |
chaque for $890.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Human Osteocrin quantitates human Osteocrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Osteocrin.
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortexSpécifications
| musclin; Ostc; Osteocrin; Ostn; Processed Osteocrin | |
| 0.55 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| OSTN | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Osteocrin | |
| 4 hrs 45 mins |
| 0.55 to 125 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 344901 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |