missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Osteocrin ELISA Kit

Catalog No. EH355RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH355RBX5 5 x 96 Tests
EH355RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH355RBX5 Supplier Invitrogen™ Supplier No. EH355RBX5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

Human Osteocrin quantitates human Osteocrin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Osteocrin.

Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex.

Specifications

Accession Number P61366
Assay Range 0.55-125 ng/mL
Assay Sensitivity 0.55 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias MUSCLIN; Ostc; osteocrin; Ostn; Processed OsteocrinMUSCLIN; Ostc; osteocrin; Ostn; Processed Osteocrin;
Gene ID (Entrez) 344901
Gene Symbol OSTN
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Osteocrin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less