Learn More
PeproTech Human IL-1 alpha ELISA Development Kit (TMB), PeproTech®
Description
900-T11K consists of: 900-T11 (capture antibody, detection antibody, and protein standard), and HRP-17UL (streptavidin HRP-conjugate). Human IL-1 alpha TMB ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinanthuman IL-1 alpha in a sandwich ELISA format. Human IL-1 alpha TMB ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells. The TMB ELISA Buffer Kit (Product # 900-T00), sold separately, is recommended to be used in conjunction with this TMB ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
Spécifications
Spécifications
| Numéro d'enregistrement | P01583 |
| Plage de l'essai | Lot-specific |
| Sensibilité d’essai | Lot-specific |
| Conjugué | HRP |
| Format de la trousse d’immunodosage | Sandwich ELISA Kit |
| Type de produit | ELISA |
| Famille de protéines | Growth Factors & Receptors |
| Espèces cibles | Human |
| À utiliser avec (équipement) | Absorbance Microplate Reader |
| Alias du gène | CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8; |
| Afficher plus de résultats |