Learn More
Invitrogen™ Human IgG1 ELISA Kit
Description
The Human Immunoglobulin G1 (Hu IgG1) ELISA quantitates Hu IgG1 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG1. Principle of the method The Human IgG1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spécifications
Spécifications
| Plage de l'essai | 15.6-1000 ng/mL |
| Sensibilité d’essai | 0.32 ng/mL |
| Contenu et stockage | Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human IgG1 HRP-Conjugate anti-human IgG monoclonal antibody human IgG1 Standard lyophilized, 2 000 ng/mL upon reconstitution Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Blue-Dye Green-Dye Adhesive Films |
| Conjugué | HRP |
| Format de la trousse d’immunodosage | Sandwich ELISA Kit |
| Type de produit | ELISA |
| Type d’échantillon | Plasma, Serum |
| Espèces cibles | Human |
| À utiliser avec (application) | ELISA |
| À utiliser avec (équipement) | Absorbance Microplate Reader |
| Afficher plus de résultats |
For Research Use Only