missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DSCAM ELISA Kit

Numéro de catalogue. EH163RBX5
Encompass
Modifier l'affichage
Click to view available options
Quantité:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
EH163RBX5 5 x 96 Tests
EH163RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. EH163RBX5 Fournisseur Invitrogen™ Code fournisseur EH163RBX5
Obtenez jusqu’à 3,000 $ de produits Extra Life Sciences avec achat
Afficher toutes les promotions

ELISA

Human DSCAM quantitates human DSCAM in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM.

This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].

Spécifications

Numéro d'enregistrement O60469
Plage de l'essai 1.4-300 ng/mL
Sensibilité d’essai 1.4 ng/mL
Conjugué HRP
Format de la trousse d’immunodosage Sandwich ELISA Kit
Type de produit ELISA
Type d’échantillon Plasma, Serum, Supernatant
Espèces cibles Human
À utiliser avec (équipement) Absorbance Microplate Reader
Alias du gène 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule;
ID du gène (Entrez) 1826
Symbole de gène DSCAM
CV intra-essai <12%, <10%
Contenus des kits Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Étiquette ou colorant Biotin
Quantité 5 x 96 Tests
Statut réglementaire RUO
Volume d’échantillon Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping condition (Condition d'expédition) Wet or Dry Ice
Conditions de stockage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Cible DSCAM
Durée du test 1 hr 20 min
Durée totale de l'essai 4 hr 45 min
Afficher plus de résultats Afficher moins de résultats