missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cystatin A ELISA Kit
Description
Human Cystatin A quantitates human Cystatin A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cystatin A.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.
Spécifications
Spécifications
| Numéro d'enregistrement | P01040 |
| Plage de l'essai | 6.5-1500 pg/mL |
| Sensibilité d’essai | 6.5 pg/mL |
| Conjugué | HRP |
| Format de la trousse d’immunodosage | Sandwich ELISA Kit |
| Type de produit | ELISA |
| Type d’échantillon | Plasma, Serum, Supernatant |
| Espèces cibles | Human |
| À utiliser avec (équipement) | Absorbance Microplate Reader |
| Alias du gène | AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFAAREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFA; |
| Afficher plus de résultats |