Learn More
Invitrogen™ Human C1q ELISA Kit
Description
Comprend
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C1q
The Human C1q ELISA quantitates Hu C1q in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu C1q. Principle of the method The Human C1q solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spécifications
Spécifications
| Numéro d'enregistrement | P02745, P02746, P02747 |
| Plage de l'essai | 0.9-60 ng/mL |
| Sensibilité d’essai | 0.08 ng/mL |
| Contenu et stockage | Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human C1q Biotin-Conjugate anti-human C1q monoclonal antibody Streptavidin-HRP Human C1q Standard lyophilized,120 ng/mL upon reconstitution Sample Diluent Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Adhesive Films |
| Conjugué | HRP |
| Format de la trousse d’immunodosage | Sandwich ELISA Kit |
| Type de produit | ELISA |
| Type d’échantillon | Plasma, Serum, Supernatant |
| Espèces cibles | Human |
| À utiliser avec (application) | ELISA |
| Afficher plus de résultats |