Learn More
G-Biosciences Hot Start Taq DNA Polymerase
Hot start Taq DNA Polymerase is used for routine PCR, qPCR, genotyping and high GC content template DNA amplification with GC enhancer.
Supplier: G-Biosciences 7861962
Description
- G-biosciences hot start Taq DNA Polymerase is chemically modified that possesses a 5' - 3' polymerase activity and a 5'-flap endonuclease activity.
- Hot start Taq DNA Polymerase reduces non-specific amplification during PCR setup and increases PCR specificity and sensitivity.
Identifiants chimiques
| qPCR | |
| High | |
| Standard | |
| DNA Polymerase |
| Liquid | |
| Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR | |
| Biotin, 6-FAM, ROX, SYBR Green, TAMRA | |
| Hot-start PCR, Multiplex PCR, Genotyping, DNA Sequencing, Next-Generation Sequencing (NGS) |
Spécifications
| qPCR | |
| High | |
| Standard | |
| DNA Polymerase | |
| 200 |
| Liquid | |
| Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR | |
| Biotin, 6-FAM, ROX, SYBR Green, TAMRA | |
| Hot-start PCR, Multiplex PCR, Genotyping, DNA Sequencing, Next-Generation Sequencing (NGS) |