missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HET-0016, 98%
$428.27
Identifiants chimiques
| CAS | 339068-25-6 |
|---|---|
| Molecular Formula | C12H18N2O |
| Synonym | N'-(4-n-Butyl-2-methylphenyl)-N-hydroxyformamidine |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AAJ67168MA
|
Thermo Scientific Chemicals
J67168MA |
10 mg |
chaque for $428.27
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 339068-25-6 | |
| C12H18N2O | |
| N'-(4-n-Butyl-2-methylphenyl)-N-hydroxyformamidine |
Spécifications
| Pale pink | |
| 339068-25-6 | |
| 206.29 | |
| HET-0016, 98% | |
| LYNOGBKNFIHKLE-UHFFFAOYSA-N | |
| N'-(4-butyl-2-methylphenyl)-N-hydroxymethanimidamide | |
| 98% |
| -30°C to -10°C | |
| C12H18N2O | |
| N'-(4-n-Butyl-2-methylphenyl)-N-hydroxyformamidine | |
| Solid | |
| CCCCC1=CC(C)=C(C=C1)N=CNO | |
| 206.29 |
Sécurité et manipulation
Recommended Storage : Store at -20°C
RUO – Research Use Only