missing translation for 'onlineSavingsMsg'
Learn More

HEPES sodium salt, ≥99%, MP Biomedicals™

Numéro de catalogue. p-4405752
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN19482825 25 g
ICN19482880 100 g
ICN19482890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. ICN19482825 Fournisseur MP Biomedicals Inc Code fournisseur 0219482825
Il en reste null

HEPES is widely used in many biochemical reactions and as a buffering agent in some cell culture media. It is used in a protein deposition technique in electron microscopy because it did not affect metal substrates

Identifiants chimiques

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Spécifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range ≥99%
Formula Weight 260.3
Chemical Name or Material HEPES sodium salt
EINECSNumber 278-169-7