missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ HEPES, Powder
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- pH 7.2 to 7.6
- Molecular Weight: 238.3
Spécifications
Spécifications
| Physical Form | Powder |
| Quantity | 1 kg |
| Packaging | Plastic Bottle |
| Sterility | Sterile |