missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Heated Tampers

These Heated Tampers are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.

Supplier:  Mopec™ BK352

 Afficher plus de versions de ce produit

Numéro de catalogue. 22445324


Il en reste null

Description

Description

  • Molded finger grips increase user comfort and remain cool when in use
  • Bayonet connection prevents disconnection while in use
  • Compatible with the Mopec™ Control Unit (BK330)
Spécifications

Identifiants chimiques

Heated Tampers
Yes
Tissue Specimens
Plastic, Metal, Wood

Spécifications

Heated Tampers
Yes
0.63 in.
Plastic, Metal, Wood
0.63 in.
Yellow
Tissue Specimens
16 mm
Medium
16 mm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions