missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ T, 60-80 mesh

Numéro de catalogue. 111004200
Modifier l'affichage
Click to view available options
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
111004200 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111004200 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 10309
Il en reste null

HayeSep™ T was used together with a thermoconductivity detector to study the solubility of trioxane in methanol at different temperatures and to measure the vapour pressure data for this binary mixture.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

Spécifications

Particle Size 60/80 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜250 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.38 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.