missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ D, 80-100 mesh

Numéro de catalogue. 111004191
Modifier l'affichage
Click to view available options
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
111004191 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111004191 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 10292
Il en reste null

HayeSep™ D in combination with Tenax TA was used to analyze polar, low-molecular weight compounds, such as methanol and ethanol, and non-polar volatile organic compounds, such as benzene and toluene in a temperature-programmed desorption combined with mass spectrometry and Fourier transform infrared spectroscopy (FTIR).

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep™ has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity.

Spécifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜795 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.33 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.