missing translation for 'onlineSavingsMsg'
Learn More

Gum arabic, MP Biomedicals™

Gum Arabic can be branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts.

$159.36 - $159.36

Identifiants chimiques

CAS 9000-01-5
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
ICN15121891
Afficher les documents
MP Biomedicals Inc
0215121891
1 kg
chaque for $159.36
Il en reste null
 
Description

Description

  • It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals.
  • In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.
Spécifications

Identifiants chimiques

9000-01-5

Spécifications

White
9000-01-5
1 kg
Gum arabic
Powder
Arabic gum, Australian gum
250,000 to 1,000,000
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'