Learn More
MilliporeSigma™ Supelco™ Graduated Screw Top Vials, PTFE/Silicone Septum
Pre-assembled, clear glass vials are supplied with a PTFE/Silicone septum
Supplier:  MilliporeSigma™ Supelco™ 27521
 Afficher plus de versions de ce produit
Numéro de catalogue. 111009448
Il en reste null
Description
Description
Spécifications
Identifiants chimiques
Black
Clear
PTFE, Silicone
100/Pk.
Open Top Cap
Screw Thread Vial
Pre-assembled
Glass
Yes
Phenolic
Autoclavable
Spécifications
Black
Clear
Glass
PTFE, Silicone
100/Pk.
Open Top Cap
82 mm
Pre-assembled
29 mm
40 mL
Yes
Phenolic
Autoclavable
Screw Thread Vial
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS