Learn More
Abcam Inc anti-Iba1, Polyclonal, Abcam™ DFS Item
Supplier:  Abcam Inc AB5076100UG
Iba1, Host Species: Goat, Target Species: Rat, Murine, Monkey, Porcine, Human, Guinea Pig, Polyclonal, Primary, 0.5mg/mL, Unconjugated, 100ug, Affinity Purified, Synthetic peptide: C-TGPPAKKAISELP, corresponding to amino acids 135-147 of Human Iba1, IgG, Cell Biology, Immunology, Neuroscience
Numéro de catalogue. NC9823189
Il en reste null
Découvrez plus d'offres spéciales