missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal dimethyl acetal, 60% aq. soln.

CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol

$96.74 - $1078.60

Identifiants chimiques

CAS 51673-84-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00134410
InChI Key OGFKTAMJLKHRAZ-UHFFFAOYSA-N
Synonym acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
PubChem CID 162650
IUPAC Name 2,2-dimethoxyacetaldehyde
SMILES COC(C=O)OC
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAL1849022
Afficher les documents
Thermo Scientific Chemicals
L1849022
100 g
chaque for $96.74
Il en reste null
 
AAL1849036
Afficher les documents
Thermo Scientific Chemicals
L1849036
500 g
chaque for $344.23
Il en reste null
 
AAL184900P
Afficher les documents
Thermo Scientific Chemicals
L184900P
2000 g
chaque for $1,078.60
Il en reste null
 
Description

Description

Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Spécifications

Identifiants chimiques

51673-84-8
104.105
OGFKTAMJLKHRAZ-UHFFFAOYSA-N
162650
COC(C=O)OC
C4H8O3
MFCD00134410
acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
2,2-dimethoxyacetaldehyde

Spécifications

51673-84-8
100°C to 102°C
C4H8O3
MFCD00134410
1850744
Fully miscible in water.
COC(C=O)OC
104.105
104.11
Glyoxal dimethyl acetal
1.15
102°C (216°F)
1.414
100 g
acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
OGFKTAMJLKHRAZ-UHFFFAOYSA-N
2,2-dimethoxyacetaldehyde
162650
60% aq. soln.
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only