missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaraldehyde, 3%, in Aqueous Phosphate Buffer, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 85.0, 11.6, 0.84, 0.52 |
| CAS Max % | 89.0, 12.07, 0.87, 0.54 |
| Percent Purity | 3% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 5.96 to 7.06 |
| Quantity | 360 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 3% |
| Show More |